NeuLog Rechargeable Battery Module
NeuLog Rechargeable Battery 202
Catalog # 470226-926
Supplier: Wards
Fournisseur #: BAT202
Jump to:
Product Details & Documents
Make your Neulog probes portable with the battery module.
- Battery module supplies power to a sensor, or chain of sensors, operating in the offline mode, and to sensors connected to an RF communication module
The battery module can be recharged by connecting it to the PC’s USB socket via the USB module’s USB cable. The battery module has a LED and a pushbutton switch. The LED indicates whether the battery is charged enough or not when pressing the pushbutton.
Caution: For educational use only.
Spécifications
- Désignation:NeuLog Rechargeable Battery 202