LITE™ Splash Shield, Unimed-Midwest
Supplier: Unimed-Midwest
Certificates
About this item
LITE™ Splash Shields deliver exceptional peripheral and forward vision, while offering reliable face and eye protection.
- Disposable, optically clear, and anti-fog treated
- Comfortable design enables all-day wear
- Fits comfortably over corrective eyewear
- Shields clip into reusable frames for biohazard protection
Delivery: Ordering Information: Shield contains 8 frames (2 each of Blue, Green, Yellow, and Violet) and 40 shields.
Frequently Bought Together