Odyssey® II Cleanroom Goggles, Sellstrom®
Supplier: Sellstrom
Certificates
About this item
Goggles with UV absorbing PC lens feature a flexible design to ensure a proper fit.
- Goggles can be autoclaved
- High temperature resistant
- Complete panoramic vision
- Ultra comfortable, soft goggle body for a snug seal
- Non-vented body offers the best protection against liquids and dust
- Synthetic Rubber Strap (Latex Free)
- Easy strap adjustment
Store in a dry, cool place away from direct sunlight (<40°C).
They can be used comfortably over most prescription glasses.
Certifications: Meets ANSI Z87.1+ High Impact Standards, CUL certified to meet CAN/CSA Z94.3, CE certified to meet EN166B.
Frequently Bought Together