T-Sue™ MicroArray Molds Kits, Simport | Avantor
T-Sue™ MicroArray Molds Kits, Simport
T-Sue™ MicroArray Molds Kits, Simport
Supplier:  Simport Scientific
Certificates
T-Sue™ MicroArray Molds Kits, Simport
T-Sue™ MicroArray Molds Kits, Simport
19 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
T-Sue™ Microarray Mold Kits enable tissues from many donor blocks to be arrayed on a single slide.
  • Can be reused hundreds of times without losing its flexibility
  • Withstands temperatures between –100 to 250°C
  • View many different samples on the same slide
The array mold is designed to be simple and easy to use
The precast T-Sue™ Microarray paraffin blocks allow users to prepare tissue arrays without needing any specialized equipment. Construct a tissue array block in minutes, simply by punching the donor tissue cores and inserting them into the pre-made paraffin recipient block. No specific technical training or experience is needed. They are ready to accept cores from a donor block, and offer a consistent core diameter. The cassette is permanently bonded to the block.
T-Sue™ punch needles are used for filling recipient tissue microarray paraffin blocks. Internal stylet operates like a retractable pen. Effortless ejection of core specimens.
Tissues can be analyzed in the same conditions enhancing the efficiency of the research. The T-Sue™ Microarray molds allow users to perform TMAs faster while giving excellent results.
Product Family Options
T-Sue™ Microarray Molds
Informations sur le produit
  • Désignation
    Availability
    Price
  • 15 Mold Cores, 4 mm Core Diameter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $1,472.72
    $1,472.72
    Pack of 1
       $1,472.72
    Supplier Number: M473-15
  • 24 Mold Cores, 2 mm Core Diameter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $1,472.72
    $1,472.72
    Pack of 1
       $1,472.72
    Supplier Number: M473-24
  • 36 Mold Cores, 3 mm Core Diameter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $1,486.59
    $1,486.59
    Pack of 1
       $1,486.59
    Supplier Number: M473-36
  • 60 Mold Cores, 2 mm Core Diameter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $1,401.26
    $1,401.26
    Pack of 1
       $1,401.26
    Supplier Number: M473-60
  • 72 Mold Cores, 1.5 mm Core Diameter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $1,381.64
    $1,381.64
    Pack of 1
       $1,381.64
    Supplier Number: M473-72
  • 150 Mold Cores, 1.5 mm Core Diameter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $1,431.44
    $1,431.44
    Pack of 1
       $1,431.44
    Supplier Number: M473-150
  • 170 Mold Cores, 1 mm Core Diameter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $1,381.64
    $1,381.64
    Pack of 1
       $1,381.64
    Supplier Number: M473-170
Precast T-Sue™ Paraffin Blocks
Informations sur le produit
  • Désignation
    Availability
    Price
  • 15 Mold Cores, 4 mm Core Diameter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 6
    Case of 6 - $220.81
    $220.81
    Case of 6
       $220.81
    Supplier Number: M473-15PC
  • 24 Mold Cores, 2 mm Core Diameter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 6
    Case of 6 - $220.81
    $220.81
    Case of 6
       $220.81
    Supplier Number: M473-24PC
  • 36 Mold Cores, 3 mm Core Diameter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 6
    Case of 6 - $220.81
    $220.81
    Case of 6
       $220.81
    Supplier Number: M473-36PC
  • 60 Mold Cores, 2 mm Core Diameter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 6
    Case of 6 - $223.93
    $223.93
    Case of 6
       $223.93
    Supplier Number: M473-60PC
  • 72 Mold Cores, 1.5 mm Core Diameter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 6
    Case of 6 - $220.81
    $220.81
    Case of 6
       $220.81
    Supplier Number: M473-72PC
  • 150 Mold Cores, 1.5 mm Core Diameter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 6
    Case of 6 - $220.81
    $220.81
    Case of 6
       $220.81
    Supplier Number: M473-150PC
  • 170 Mold Cores, 1 mm Core Diameter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 6
    Case of 6 - $220.81
    $220.81
    Case of 6
       $220.81
    Supplier Number: M473-170PC
T-Sue™ Punch Needles
Informations sur le produit
  • Désignation
    Availability
    Price
  • 1 mm Diameter, Magenta
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 4
    Pack of 4 - $100.40
    $100.40
    Pack of 4
       $100.40
    Supplier Number: M473-P10MA
  • 1.5 mm Diameter, Yellow
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 4
    Pack of 4 - $107.01
    $107.01
    Pack of 4
       $107.01
    Supplier Number: M473-P15Y
  • 2 mm Diameter, Red
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 4
    Pack of 4 - $100.40
    $100.40
    Pack of 4
       $100.40
    Supplier Number: M473-P20R
  • 3 mm Diameter, Pink
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 4
    Pack of 4 - $105.98
    $105.98
    Pack of 4
       $105.98
    Supplier Number: M473-P30P
  • 4 mm Diameter, Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 4
    Pack of 4 - $105.98
    $105.98
    Pack of 4
       $105.98
    Supplier Number: M473-P40B