96-Well Blocks, Zymo Research
Fournisseur: Zymo Research
Certificats
À propos de cet article
2 mL deep-well block for processing and mixing of samples.
- Conforms to laboratory standards
96-Well Block features durable, clear polypropylene construction. Each has a level footprint and conforms to laboratory standards. Capacity is 2mL per round bottom well.
Frequently Bought Together