Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:2 mg
- Storage conditions:–20 °C
- Gene ID:P0CG47
- Protein synonyms:Ubiquitin B|UBB
- Protein/peptide name:Ubiquitin
- Formulation:Lyophilized from a solution in deionized water.
- Shipping temperature:-20 °C, Blue Ice
- Cat. no.:102980-888
- Supplier No.:AG40T0464M002
Specifications
About this item
Ubiquitin is a 76 amino acid (aa) protein that is ubiquitously expressed in all eukaryotic organisms. ubiquitin is highly conserved with 96% aa sequence identity shared between human and yeast ubiquitin, and 100% aa sequence identity shared between human and mouse ubiquitin. In mammals, four ubiquitin genes encode for two ubiquitin-ribosomal fusion proteins and two poly-ubiquitin proteins. Cleavage of the ubiquitin precursors by deubiquitinating enzymes gives rise to identical ubiquitin monomers each with a predicted molecular weight of 8.6 kDa. Conjugation of ubiquitin to target proteins involves the formation of an isopeptide bond between the C-terminal glycine residue of ubiquitin and a lysine residue in the target protein. This process of conjugation, referred to as ubiquitination or ubiquitylation, is a multi-step process that requires three enzymes: a ubiquitin-activating (E1) enzyme, a ubiquitin-conjugating (E2) enzyme, and a ubiquitin ligase (E3). ubiquitination is classically recognized as a mechanism to target proteins for degradation and as a result, ubiquitin was originally named ATP-dependent Proteolysis Factor 1 (APF-1). In addition to protein degradation, ubiquitination has been shown to mediate a variety of biological processes such as signal transduction, endocytosis, and post-endocytic sorting.