YES Medium Plates, Teknova
YES Medium Plates, Teknova
Supplier:   Teknova
YES Medium Plates, Teknova
Supplier:  Teknova
Certificates
YES Medium Plates, Teknova
2 Options Available Below
Product Details & Documents
For the propagation and maintenance of yeasts for use in molecular biology.
  • 20 plates
  • Sterile
Contains 0.5% Yeast extract, 3.0% glucose, 0.0225% adenine, 0.0225% histidine, 0.0225% leucine, 0.0225% uracil, 0.0225% lysine, and 2.0% agar.
Ambient shipping conditions. Refrigerate to store.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • YES medium plates
    Log in for availability
    Log in for availability
    Each
    Each - $212.23
    $212.23
    Each
       $212.23
    Minimum order quantity is 4
    Supplier Number: Y3005
    Specifications:
    New Product
    New Product
  • YES medium plates
    Log in for availability
    Log in for availability
    Pack of 20
    Pack of 20 - $828.23
    $828.23
    Pack of 20
       $828.23
    Minimum order quantity is 8
    Supplier Number: Y3005