YES Medium Plates, Teknova
Supplier: Teknova
Jump to:
About this item
For the propagation and maintenance of yeasts for use in molecular biology.
- 20 plates
- Sterile
Contains 0.5% Yeast extract, 3.0% glucose, 0.0225% adenine, 0.0225% histidine, 0.0225% leucine, 0.0225% uracil, 0.0225% lysine, and 2.0% agar.
Ambient shipping conditions. Refrigerate to store.
2 Options Available Below
Product Details & Documents
For the propagation and maintenance of yeasts for use in molecular biology.
- 20 plates
- Sterile
Contains 0.5% Yeast extract, 3.0% glucose, 0.0225% adenine, 0.0225% histidine, 0.0225% leucine, 0.0225% uracil, 0.0225% lysine, and 2.0% agar.
Ambient shipping conditions. Refrigerate to store.