Ward's® Stackable Electrophoresis Power Supply
Ward’s Stackable Power Supply
Catalog # 470144-886
Supplier: Labnet International
Fournisseur #: E0303-WAR
Jump to:
Product Details & Documents
Versatile power supply useful for DNA/RNA or Protein electrophoresis.
- Dependable performance, simple to program
- Pause/resume function and timer with alarm
- Advanced safety features
Safety features incorporated into the systems include automatic crossover, no load detection, sudden load change and over voltage protection. The units can also be programmed to restart in the event of a power disruption.
Spécifications
- Désignation:Ward’s Stackable Power Supply
- Cat. No.:470144-886
- Timer:0 - 99:59 h
- Output voltage range/increments:2 - 300/1 V
- Output current range/increments:4 - 500/1 mA
- Electrical:120 V