KIMAX® Centrifuge Tubes, Conical-Bottom, with Flathead PTFE Stoppers, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
À propos de cet article
Standard Taper KIMAX® tube is made with heavier-walled tubing to withstand higher centrifugation speeds.
- Manufactured from 33 expansion, low extractable borosilicate glass
- Conical bottom
- Color-coded flathead PTFE stopper
Certifications: Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Frequently Bought Together