SEM Clip Specimen Mounts, Electron Microscopy Sciences
SEM Clip Specimen Mounts, Electron Microscopy Sciences
SEM Clip Specimen Mounts, Electron Microscopy Sciences
Certificates
SEM Clip Specimen Mounts, Electron Microscopy Sciences
SEM Clip Specimen Mounts, Electron Microscopy Sciences
11 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Specially designed for easy, quick and clean mounting of any type of thin specimen on specimen stubs.
  • Available with single and multiple clips
  • Ideal for holding silicon chips, paper, wire, threads, thin films, sheet metal, and more
  • SEM clip mounts with multiple clips can hold larger samples or multiple smaller samples
These spring-loaded clips accommodate samples of up to 2 mm thickness, samples are held securely and allow for easy change of samples. Adhesives are not required, eliminating possible outgassing issues and saving time.
Product Family Options
Informations sur le produit
  • Désignation
    Ø
    Availability
    Price
  • SEM Clip, 18 mm Pin Mount, 6 mm pin height, 1 clip
    18 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $85.76
    $85.76
    Each
       $85.76
    Supplier Number: 75923-11
    Spécifications:
    Marketsource
    Marketsource
  • SEM Clip, 25 mm Pin Mount, 1 clip
    25 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $69.29
    $69.29
    Each
       $69.29
    Supplier Number: 75923-12
  • SEM Clip, 25 mm Pin Mount, 2 clips
    25 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $86.54
    $86.54
    Each
       $86.54
    Supplier Number: 75923-13
  • SEM Clip, 25 mm Pin Mount, for ZEISS/LEO SEM/FIBs 6 mm pin height, 1 clip
    25 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $67.34
    $67.34
    Each
       $67.34
    Supplier Number: 75923-15
  • SEM Clip, 25 mm Pin Mount, for ZEISS/LEO SEM/FIBs 6 mm pin height, 3 clips
    25 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $103.15
    $103.15
    Each
       $103.15
    Supplier Number: 75923-17
  • SEM Clip, 32 mm Pin Mount, 1 clip
    38 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $69.29
    $69.29
    Each
       $69.29
    Supplier Number: 75923-18
  • SEM Clip, 32 mm Pin Mount, 2 clips
    38 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $82.51
    $82.51
    Each
       $82.51
    Supplier Number: 75923-19
    Spécifications:
    Marketsource
    Marketsource
  • SEM Clip, 32 mm Pin Mount, 3 clips
    38 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $103.15
    $103.15
    Each
       $103.15
    Supplier Number: 75923-20
  • SEM Clip, 38 mm Pin Mount, 1 clip
    38 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $82.65
    $82.65
    Each
       $82.65
    Supplier Number: 75923-21
  • SEM Clip, 38 mm Pin Mount, 2 clips
    38 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $100.72
    $100.72
    Each
       $100.72
    Supplier Number: 75923-22
  • SEM Clip, 38 mm Pin Mount, 3 clips
    38 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $121.26
    $121.26
    Each
       $121.26
    Supplier Number: 75923-23