MacroTubes, 5 ml
MacroTubes, 5 ml
MacroTubes, 5 ml
MacroTubes, 5 ml
The perfect tubes for intermediate volumes.
  • Uniform wall thickness
  • Appropriate for storage from –86 to 100 °C
  • Color-code samples for easy identification
  • RNAse-, DNAse-, Pyrogen-, and Cytotoxin-free
  • Snap-cap or screw-cap available
Made of maximum clarity, low-binding virgin polypropylene, these tubes are sized to be compatible with all standard equipment.
They're a valuable resource for boiling and for sample preservation.
Product Family Options
Snap Cap Tubes
  • Désignation
    Dimensions
    Maximum RCF
    Packaging
    Sterility
  • Natural Colored Tubes
    16 x 55 mm
    25000×g
    4 Bags of 50 Tubes
    Non sterile
    Case of 200
    Case of 200 - $105.90
    $105.90
    Case of 200
       $105.90
     : C2500_WARDS
  • Natural Colored Tubes
    16 x 55 mm
    25000×g
    10 Bags of 20 Tubes
    Sterile
    Case of 200
    Case of 200 - $100.20
    $100.20
    Case of 200
       $100.20
     : C2520_WARDS
  • Red Colored Tubes
    16 x 55 mm
    25000×g
    4 Bags of 50 Tubes
    Non sterile
    Case of 200
    Case of 200 - $98.45
    $98.45
    Case of 200
       $98.45
     : C2500-R_WARDS
  • Opaque black Colored Tubes
    16 x 55 mm
    25000×g
    4 Bags of 50 Tubes
    Non sterile
    Case of 200
    Case of 200 - $98.45
    $98.45
    Case of 200
       $98.45
     : C2500-OB_WARDS
  • Yellow Colored Tubes
    16 x 55 mm
    25000×g
    4 Bags of 50 Tubes
    Non sterile
    Case of 200
    Case of 200 - $98.45
    $98.45
    Case of 200
       $98.45
     : C2500-Y_WARDS
  • Green Colored Tubes
    16 x 55 mm
    25000×g
    4 Bags of 50 Tubes
    Non sterile
    Case of 200
    Case of 200 - $98.45
    $98.45
    Case of 200
       $98.45
     : C2500-G
  • Blue Colored Tubes
    16 x 55 mm
    25000×g
    4 Bags of 50 Tubes
    Non sterile
    Case of 200
    Case of 200 - $98.45
    $98.45
    Case of 200
       $98.45
     : C2500-B_WARDS
  • Assorted Colors
    16 x 55 mm
    25000×g
    4 Bags of 50 Tubes
    Non sterile
    Case of 200
    Case of 200 - $85.30
    $85.30
    Case of 200
       $85.30
     : C2500-AST_WARD
Screw Cap Tubes
  • Désignation
    Dimensions
    Maximum RCF
    Packaging
    Sterility
  • Natural Colored Tubes
    16 x 64 mm
    25000×g
    5 Bags of 100 Tubes
    Non sterile
    Case of 500
    Case of 500 - $236.10
    $236.10
    Case of 500
       $236.10
     : C2530_WARDS
  • Natural Colored Tubes
    16 x 64 mm
    25000×g
    10 racks of 50 Tubes
    Sterile
    Case of 500
    Case of 500 - $261.60
    $261.60
    Case of 500
       $261.60
     : C2540_WARDS
Optional Accessories
  • Désignation
    Dimensions
    Maximum RCF
    Packaging
    Sterility
  • Cryobox Storage Box for 25 Tubes
    5.25 x 5.25 x 3"
    Pack of 5
    Pack of 5 - $172.20
    $172.20
    Pack of 5
       $172.20
  • Workstation Rack Tube Holder for 50 Tubes
    9.5 x 5 x 2"
    Each
    Each - $119.75
    $119.75
    Each
       $119.75
  • Adapter for 15mL Centrifuge
    4 per Pack
    Pack of 4
    Pack of 4 - $98.45
    $98.45
    Pack of 4
       $98.45
  • Lock Clip for Snap Cap Tubes
    20 per Pack
    Pack of 20
    Pack of 20 - $61.60
    $61.60
    Pack of 20
       $61.60