Polyethylene Transfer Pipettes
Polyethylene Transfer Pipettes
Polyethylene Transfer Pipettes
Supplier:  VWR International
Certificates
Polyethylene Transfer Pipettes
Polyethylene Transfer Pipettes
16 Options Available Below
Product Details & Documents
Disposable transfer pipettes for liquid handling.
  • Transparent, non-toxic, and inert
  • Precise, uniform drop size
  • Shatter-proof and unbreakable
  • Can be used with liquid nitrogen
  • Heat sealable for liquid storage
Unbreakable, one-piece pipettes eliminate the need for rubber bulbs, broken glass, and exposure to infected material.
Ideal for microbiology, blood banking, hematology, immunology, and urinalysis. Pipettes can also be heat sealed for airtight sample storage.
Packaging: Available packaging includes nonsterile bulk, sterile individually wrapped, or sterile packs of 20.
Documents
Product Family Options
Individually Wrapped - Sterile
Informations sur le produit
  • Désignation
    Overall Length
    Drops per mL
    Graduation
    Bulb Draw
    Availability
    Price
  • 1.5 ml Small Bulb Extended Tip
    104 mm
    50
    Non-Graduated
    1.0 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $160.70
    $160.70
    Case of 500
       $160.70
  • 3.0 ml Short Bulb
    138 mm
    22
    Graduated to 1 ml
    1.5 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $136.35
    $136.35
    Case of 500
       $136.35
  • 4.0 ml
    150 mm
    28
    Non-Graduated
    3.5 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $127.35
    $127.35
    Case of 500
       $127.35
  • 5.0 ml Large Bulb
    150 mm
    22
    Graduated
    3.5 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $137.40
    $137.40
    Case of 500
       $137.40
  • 5.0 ml Large Bulb
    160 mm
    25
    Graduated
    3.5 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $136.35
    $136.35
    Case of 500
       $136.35
  • 7.0 ml Large Bulb Extended Tip
    150 mm
    66
    Non-Graduated
    6.0 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $213.20
    $213.20
    Case of 500
       $213.20
  • 7.0 mL Large Bulb
    160 mm
    21
    Graduated to 3 ml
    3.5 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $136.35
    $136.35
    Case of 500
       $136.35
20 Pack - Sterile
Informations sur le produit
  • Désignation
    Overall Length
    Drops per mL
    Graduation
    Bulb Draw
    Availability
    Price
  • 4.0 ml
    150 mm
    28
    Non-Graduated
    3.5 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $103.10
    $103.10
    Case of 500
       $103.10
  • 5.0 Large Bulb
    150 mm
    22
    Graduated
    3.5 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $100.90
    $100.90
    Case of 500
       $100.90
  • 7.0 ml Large Bulb
    160 mm
    21
    Graduated to 3 ml
    3.5 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $95.90
    $95.90
    Case of 500
       $95.90
Bulk Bag - Nonsterile
Informations sur le produit
  • Désignation
    Overall Length
    Drops per mL
    Graduation
    Bulb Draw
    Availability
    Price
  • 1.5 ml Small Bulb Extended Tip
    104 mm
    50
    Non-Graduated
    1.0 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $111.75
    $111.75
    Case of 500
       $111.75
  • 3.0 ml Short Bulb
    138 mm
    22
    Graduated to 1 ml
    1.5 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 250
    Case of 250 - $42.45
    $42.45
    Case of 250
       $42.45
  • 5.0 ml Large Bulb
    150 mm
    22
    Graduated
    3.5 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 250
    Case of 250 - $42.45
    $42.45
    Case of 250
       $42.45
  • 5.0 ml Large Bulb
    160 mm
    25
    Graduated to 1 ml
    3.5 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 250
    Case of 250 - $42.45
    $42.45
    Case of 250
       $42.45
  • 7.0 ml Large Bulb
    160 mm
    21
    Graduated to 3 ml
    3.5 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 250
    Case of 250 - $42.45
    $42.45
    Case of 250
       $42.45
  • 7.0 ml Large Bulb Extended Tip
    150 mm
    66
    Non-Graduated
    6.0 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 400
    Case of 400 - $111.75
    $111.75
    Case of 400
       $111.75