Rough Surface Polyester Labels, Type B3, ToughBond®, Brady
Rough surface polyester labels, B-432, Polyester, Clear, BBP®35; BBP®37, 4", 36.6 m (120 ft.)
Catalog # 10017-992
Supplier: Brady Worldwide
Fournisseur #: 145292
Rough Surface Polyester Labels, Type B3, ToughBond®, Brady
Catalog # 10017-992
Supplier: Brady Worldwide
Jump to:
Product Details & Documents
Rough Surface Polyester (B-432) labels are ideal for electronic PCB and component identification, barcode labels, rating plates, and solar panel identification.
- Made from durable BradyBondz™ glossy white polyester with permanent acrylic adhesive, labels are chemical, oil, and water resistant
- UL Recognized for outdoor use on glass, thermoset polyester plastic, and polyvinyl fluoride plastic surfaces to support solar panel identification
- Enables printers with XY cutters to cut custom shapes or multiple labels out of a single continuous roll
Clear polyester material with aggressive permanent adhesive for labeling products and general identification. Best for application to smooth and textured surfaces indoors. Cut multiple labels out of a single 4" wide material using BBP®37 Printer.
Certifications: RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; CSA accepted
Spécifications
- Pour l'utilisation avec:BBP®35; BBP®37
- Color:Clear
- Désignation:Rough surface polyester labels, B-432
- Matière:Polyester
- Roll Length:36.6 m (120 ft.)
- Width:4"