Hazardous Material D.O.T. Vehicle Placards: Explosive, Brady
Supplier: Brady Worldwide
Certificates
About this item
D.O.T
- Placards meet D.O.T. requirements 49CFR Part 172
- Self sticking vehicle placards may be applied directly to a vehicle
- High gloss, durable, flexible vinyl with a permanent cold temperature pressure sensitive acrylic adhesive
- Good chemical, oil, and water resistance
B-946 vinyl (SSV) placards may be applied directly to a vehicle, and will provide over five years of outdoor durability. All placards come with square corners and no holes. Custom placards are also available upon request.
Vehicle placards help ensure that hazardous material shipments reach their destinations without costly delays or rejections. All placards have square corners, no holes, and no headers.
Certifications: ANSI Z53.1-1967. ANSI Z535-2006. OSHA 1910.145.
Frequently Bought Together