BCR® 3 Small Flat Mop Head Frame, Berkshire
Catalogue # 76003-526
Fournisseur: Berkshire
Spécifications
- Description:BCR® 3 Small Flat Mop Head Frame
- Dimensions:28×9 cm (11×3.5")
- Material:Aluminum, anodized
- Cat. no.:76003-526
Spécifications
À propos de cet article
Berkshire’s Flat Mop Head is light and easy to maneuver, even with wet wipers attached. The stainless steel springs or plastic eyes hold multiple mop wipers.
- Clean, light, and durable
- High quality anodized aluminum
- Autoclavable (plastic eyes are not autoclavable)
- Plastic eyes for wiper attachment
Efficient cleaning of floors, walls, and other large surfaces is critical to the overall cleanliness of the production environment. BCR® Mop Systems replicate the most effective way to clean a surface - wiping. Berkshire’s mop systems are easy to use, highly effective, and quick.
The system is very flexible, combining a wide range of alternative Mop Wipers designed for particular levels of cleanliness with a range of Flat Mop Heads.
Certifications: Designed for use in ISO class 3 to 8 environments. Cleanroom class 1 to 100000.