SiliaBond® C18 WPD, Wide Pore, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
Certificates
About this item
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
- Great variety of functional groups available
- Fast Kinetics
- Compatible with all solvents
- Excellent Mechanical and Thermal Stability
- Controlled Loading
- Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together