Scale-Up Series Cylindrical Flasks, Ace Glass
Supplier: Ace Glass
Certificates
About this item
Scale-Up Series cylindrical flasks with an Easy Drain glass valve are graduated on both sides.
- High-quality borosilicate glass
- Flask is graduated on both sides for either left or right-hand orientation
- Flasks scale up from 100 mL to 6000 mL, and their geometry translates into ACE Kilo Scale flasks
Cylindrical flasks with an Easy Drain valve which is designed to prevent accidental removal when opening, and incorporates a compression tube angled side-arm for maximum unobstructed drainage. Flasks are graduated on both sides to allow for orientation in either direction. Flasks are sized for bench-top work, but their geometry translates (scale-up) to ACE Kilo Scale flasks.
Frequently Bought Together