SDS-Page Running Gel Buffers, Rockland
Catalogue # CAMB-018
Fournisseur: Rockland Immunochemical
Numéro CAS:
Spécifications
- Description:SDS-PAGE Sample Buffer without DTT or [beta]MeOH, 2X
- Size:100 mL
- Cat. no.:CAMB-018
- pH:6.8
- Supplier No.:MB-018
Spécifications
À propos de cet article
SDS-PAGE, sodium dodecyl sulfate polyacrylamide gel electrophoresis, describes a technique widely used in biochemistry to separate proteins according to their electrophoretic mobility (a function of the length of a polypeptide chain and its charge) and no other physical feature
SDS is an anionic detergent applied to protein sample to linearize proteins and to impart a negative charge to linearized proteins
Caution: This product is for research use only and is not intended for therapeutic or diagnostic applications.