McFarland Standard, Pro-Lab Diagnostic
Supplier: PRO-LAB DIAGNOSTICS INC CA
Certificates
About this item
The McFarland Standards are intended for adjusting densities of bacterial suspensions and have a shelf life of up to two years from the date of manufacture.
Delivery: All standards are supplied in a protective carrying case.
Frequently Bought Together