Order Entry
Canada
ContactUsLinkComponent
Puritan® Fecal Opti-Swab® Collection & Transport System, Puritan Medical Products
Puritan® Fecal Opti-Swab® Collection & Transport System, Puritan Medical Products
Catalog # 75999-268
CAS Number:  
undefined
Puritan® Fecal Opti-Swab® Collection & Transport System, Puritan Medical Products
Catalog # 75999-268
Supplier Number:  CB-206
CAS Number:  

Specifications

  • Description:
    Cary-Blair Medium Filled Vial with Elongated HydraFlock Swab
  • Size:
    12 x 80 mm
  • Cat. no.:
    75999-268
  • Fill Volume:
    2 ml

Specifications

About this item

For the collection, transport, and preservation of enteric bacteria.

  • Ready-to-use, complete transport kit
  • Advance kit design
  • Self-centering cap
  • High-performance swab
  • Handle has a molded breakpoint
  • Shelf life - 18 months

The advance kit design is easy to use and reliabls. Self-centering cap is leakproof and facilitates easy swab removal and disposal. High-performance swab features a patented HydraFlock tip. Handle has a molded breakpoint.

Certifications: Made in USA.
Ordering information: Products are final sale.