KIMBLE® Glass Threaded Microscale Connecting Adapters with Outer Standard Tapered Joints, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Connecting adapters allow interfacing of Standard Taper ground jointed glassware with threaded microscale components.
- Connects standard taper outer ground joint to GPI thread of inner ground joint
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Frequently Bought Together