White Urea Closures with Cork-Backed Aluminium Foil Liners, Kimble Chase, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
À propos de cet article
Closure is made from white urea and features a cork-backed aluminum foil liner.
- Top is suitable for marking
- Ideal for scintillation vials
- Closure is not autoclavable
Frequently Bought Together