Puritan® Cary-Blair Transport Medium, Puritan Medical Products
Fournisseur: Puritan Medical Products
Certificats
À propos de cet article
Cary-Blair transport medium for the preservation of fecal specimens containing enteric pathogenic bacteria.
- 2mL or 5mL Cary-Blair Transport Medium
- Intended for use in the preservation of specimens containing enteric bacteria
- Orange cap
Cary-Blair Transport Medium is intended for use in the preservation of clinical fecal and rectal specimens containing enteric bacteria from the collection site to the testing laboratory for examination and culture. For optimum performance, store at 2-25°C.
Certifications: Made in USA.
Ordering information: Products are final sale.
Frequently Bought Together