ScienceLine pH Combination Electrodes with Temperature Sensor, SI Analytics
Supplier: SI Analytics
ScienceLine pH Combination Electrodes with Temperature Sensor, SI Analytics
Supplier: SI Analytics
Certificates
À propos de cet article
These pH combination electrode have MMR tubes with a diameter of 3 mm and a length of 180 mm
They feature round bottom flask with NS 14.5 and NS 7.5 standard taper.
1 Options Available Below
Product Details & Documents
These pH combination electrode have MMR tubes with a diameter of 3 mm and a length of 180 mm
They feature round bottom flask with NS 14.5 and NS 7.5 standard taper.
Specifications for Product Family
Spécifications
- Catalog No:
- 97042-062
- 97042-126
- 97042-076
- 97042-134
- 97042-138
- 97042-142
- 97042-350
- 97042-430
- 97042-072
- 97042-080
- 97042-128
- 97042-074
- 97042-078
- 97042-136
- 97042-140
- Thermistor:
- Pt 1000
- Pt 1000
- Pt 1000
- Pt 1000
- Pt 1000
- Pt 1000
- Pt 1000
- Pt 1000
- Pt 1000
- Pt 1000
- Pt 1000
- Pt 1000
- Pt 1000
- Pt 1000
- Pt 1000
- Shape:
- Sphere
- Sphere
- Sphere
- Sphere
- Sphere
- Sphere
- Sphere
- Sphere
- Sphere
- Sphere
- Sphere
- Sphere
- Sphere
- Sphere
- Sphere
- Zero-point:
- pH 7.0 ± 0.3
- pH 7.0 ± 0.3
- pH 7.0 ± 0.3
- pH 7.0 ± 0.3
- pH 7.0 ± 0.3
- pH 7.0 ± 0.3
- pH 7.0 ± 0.3
- pH 7.0 ± 0.3
- pH 7.0 ± 0.3
- pH 7.0 ± 0.3
- pH 7.0 ± 0.3
- pH 7.0 ± 0.3
- pH 7.0 ± 0.3
- pH 7.0 ± 0.3
- pH 7.0 ± 0.3
- Reference system:
- Silamid®
- Silamid®
- Silamid®
- Silamid®
- Silamid®
- Silamid®
- Silamid®
- Silamid®
- Silamid®
- Silamid®
- Silamid®
- Silamid®
- Silamid®
- Silamid®
- Silamid®
- Shaft material:
- Glass
- Glass
- Glass
- Glass
- Glass
- Glass
- Glass
- Glass
- Glass
- Glass
- Glass
- Glass
- Glass
- Glass
- Glass
Spécifications
Product Family Options
Informations sur le produit
- TypeØLongueurCable LengthConnecteurElectrolytePlage de températurepH RangeAvailabilityPrice
- N 1052 A12 mm120 mm1 m (fixed length)DIN and 4 mmKCl 3 mol/L‒5 ... +100 °C0 - 14Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.97042-062ThermistorPt 1000ShapeSphereZero-pointpH 7.0 ± 0.3Reference systemSilamid®Shaft materialGlass - A 7780 1M-DIN-ID12 mm120 mm1 m (fixed length)DIN and 4 mmKCl 3 mol/L‒5 ... +80 °C0 - 14Log in pour la disponibilitéEach$2,276.27Supplier Number: 285130200Spécifications:
More
Specifications
Cat. No.97042-126ThermistorPt 1000ShapeSphereZero-pointpH 7.0 ± 0.3Reference systemSilamid®Shaft materialGlass - N 1051 A12 mm170 mm1 m (fixed length)DIN and 4 mmKCl 3 mol/L‒5 ... +100 °C0 - 14Discontinued-Supplier Number: 285100510Spécifications:
More
Specifications
Cat. No.97042-076ThermistorPt 1000ShapeSphereZero-pointpH 7.0 ± 0.3Reference systemSilamid®Shaft materialGlass - A 161 1M-DIN-ID12 mm170 mm1 m (fixed length)DIN and 4 mmKCl 3 mol/L‒5 ... +100 °C0 - 14Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.97042-134ThermistorPt 1000ShapeSphereZero-pointpH 7.0 ± 0.3Reference systemSilamid®Shaft materialGlass - H 161 1M-DIN-ID12 mm170 mm1 m (fixed length)DIN and 4 mmKCl 3 mol/L+10 ... +100 °C0 - 14Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.97042-138ThermistorPt 1000ShapeSphereZero-pointpH 7.0 ± 0.3Reference systemSilamid®Shaft materialGlass - A 164 1M-DIN-ID12 mm170 mm1 m (fixed length)DIN and 4 mmKCl 3 mol/L‒5 ... +100 °C0 - 14Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.97042-142ThermistorPt 1000ShapeSphereZero-pointpH 7.0 ± 0.3Reference systemSilamid®Shaft materialGlass - N 1043 A12 mm320 mm1 m (fixed length)DIN and 4 mmKCl 3 mol/L‒5...+100 °C0 - 14Discontinued-Supplier Number: 285093009Spécifications:
More
Specifications
Cat. No.97042-350ThermistorPt 1000ShapeSphereZero-pointpH 7.0 ± 0.3Reference systemSilamid®Shaft materialGlass - N 1041 A-60012 mm600 mm1 m (fixed length)DIN and 4 mmKCl 3 mol/L‒5...+100 °C0 - 14Discontinued-Supplier Number: 285093111Spécifications:
More
Specifications
Cat. No.97042-430ThermistorPt 1000ShapeSphereZero-pointpH 7.0 ± 0.3Reference systemSilamid®Shaft materialGlass - N 1052 BNC12 mm120 mm1 m (fixed length)BNC and 4 mmKCl 3 mol/L‒5 ... +100 °C0 - 14Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.97042-072ThermistorPt 1000ShapeSphereZero-pointpH 7.0 ± 0.3Reference systemSilamid®Shaft materialGlass - N 1042 BNC12 mm120 mm1 m (fixed length)BNC and 4 mmKCl 3 mol/L‒5 ... +100 °C0 - 14Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.97042-080ThermistorPt 1000ShapeSphereZero-pointpH 7.0 ± 0.3Reference systemSilamid®Shaft materialGlass - A 7780 1M-BNC-ID12 mm120 mm1 m (fixed length)BNC and 4 mmKCl 3 mol/L‒5 ... +80 °C0 - 14Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.97042-128ThermistorPt 1000ShapeSphereZero-pointpH 7.0 ± 0.3Reference systemSilamid®Shaft materialGlass - N 1051 BNC12 mm170 mm1 m (fixed length)BNC and 4 mmKCl 3 mol/L‒5 ... +100 °C0 - 14Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.97042-074ThermistorPt 1000ShapeSphereZero-pointpH 7.0 ± 0.3Reference systemSilamid®Shaft materialGlass - N 1041 BNC12 mm170 mm1 m (fixed length)BNC and 4 mmKCl 3 mol/L‒5 ... +100 °C0 - 14Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.97042-078ThermistorPt 1000ShapeSphereZero-pointpH 7.0 ± 0.3Reference systemSilamid®Shaft materialGlass - A 161 1M-BNC-ID12 mm170 mm1 m (fixed length)BNC and 4 mmKCl 3 mol/L‒5 ... +100 °C0 - 14Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.97042-136ThermistorPt 1000ShapeSphereZero-pointpH 7.0 ± 0.3Reference systemSilamid®Shaft materialGlass - H 161 1M-BNC-ID12 mm170 mm1 m (fixed length)BNC and 4 mmKCl 3 mol/L+10 ... +100 °C0 - 14Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.97042-140ThermistorPt 1000ShapeSphereZero-pointpH 7.0 ± 0.3Reference systemSilamid®Shaft materialGlass