ContactUsLinkComponent
Hydrion Insta-Chek® pH Paper, Micro Essential Laboratory
Hydrion Insta-Chek® pH Paper, Micro Essential Laboratory
Catalog # 89370-900
Supplier:  Micro Essential
undefined
Hydrion Insta-Chek® pH Paper, Micro Essential Laboratory
Catalog # 89370-900
Supplier:  Micro Essential
Supplier Number:  93
Specifications
  • Description:
    Insta-Check 0–13 pH papers, 0-13 pH, 7/32" x 15', Each
  • Cat. no.:
    89370-900
  • pH Range:
    1.0 - 13.0
  • Supplier No.:
    93
Specifications
About this item
Hydrion Insta-Chek® pH paper, the ultimate in wide range, general purpose pH papers has a distinct color match at each full pH unit.
  • A direct reading pH paper with the widest range available in a single paper
  • A simple, single color match for all pH values
  • Distinct and easily matched color separations
  • Accurate, reproducible readings with an immediate response
Delivery: Each carton contains 10 kits, each consisting of a 15-foot roll of test paper and matching color chart. Each kit provides enough paper for approximately 100 tests. A carton of 10 provides approximately 1,000 tests.