National Basic Sensor® Temperature Sensor with Dual Sensor Probe, Ace Glass | Avantor
National Basic Sensor® Temperature Sensor with Dual Sensor Probe, Ace Glass
Supplier:   Ace Glass
National Basic Sensor® Temperature Sensor with Dual Sensor Probe, Ace Glass
Supplier:  Ace Glass
Certificates
National Basic Sensor® Temperature Sensor with Dual Sensor Probe, Ace Glass
National Basic Sensor® Temperature Sensor with Dual Sensor Probe, Ace Glass
2 Options Available Below
Product Details & Documents
Temperature sensor with two 72-inch SMP leads, for use with limit controllers, or for sending data to two different temperature devices simultaneously.
  • Maximum sheath temperature: 550 °C
  • 72-inch SMP leads
Inconel sheaths offer corrosion resistance and good high-temperature strength.
Product Family Options
Informations sur le produit
  • Désignation
    Ø
    Longueur
    Cable Length
    Connecteur
    Availability
    Price
  • K Dual Sensor Probe with Inconel Sheath
    1/4"
    12"
    72"
    SMP
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $648.59
    $648.59
    Prix promotionnel pour les nouveaux clients
    Each
       $648.59
    Supplier Number: 12144-22
  • T Dual Sensor Probe with 316SS Sheath
    1/4"
    12"
    72"
    SMP
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $645.29
    $645.29
    Prix promotionnel pour les nouveaux clients
    Each
       $645.29
    Supplier Number: 12144-24