ContactUsLinkComponent
Ward's® Essentials Acids, Bases, and the pH Scale Lab Kit
Ward's® Essentials Acids, Bases, and the pH Scale Lab Kit
  470225-526
undefined
Ward's® Essentials Acids, Bases, and the pH Scale Lab Kit
  470225-526
 :  AD-WE2507-KIT
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Acids, Bases, and the pH Scale Lab Kit
  • No. of Lab Groups:
    15
  • Cat. no.:
    470225-526
  • Supplier No.:
    AD-WE2507-KIT
Examine the effects of acids and bases on chemical pH indicators.
  • Hands on lab activity
  • Kit aligned to Next Generation Science Standards MS-PS1-2
  • Instructions and materials included for 15 groups
  • Middle school
In this lab, students will examine the effects of acids and bases on several chemical pH indicators, determine the pH of several common household materials and use the knowledge they have gained to determine the composition of four unknown solutions.