IntelliCAL PHC108 Puncture pH Combination Electrode, Hach | Avantor
IntelliCAL PHC108 Puncture pH Combination Electrode, Hach
Supplier:   Hach
IntelliCAL PHC108 Puncture pH Combination Electrode, Hach
Supplier:  Hach
Certificates
IntelliCAL PHC108 Puncture pH Combination Electrode, Hach
IntelliCAL PHC108 Puncture pH Combination Electrode, Hach
1 Options Available Below
Product Details & Documents
IntelliCAL™ PHC108 is a robust digital puncture pH electrode with stainless steel body, Ag/AgCl reference system and built-in temperature sensor.
  • Compatible with digital Hach meters only (e.g. HQd meters)
  • Digital electrode stores time and date stamp for each measurement, operator and sample ID, calibration history, parameter, and probe serial number
  • Can be moved between meters without the need to re-calibrate or re-enter measurement settings
  • Ideal for a multi-user environment with multiple HQD series laboratory and portable meters
  • Ultimate traceability in measurement history
  • Non refillable
Ideal for measuring in semi solid samples such as sludge, meats and cheeses.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 10146-090
  • CA10146-090
  • Junction:
  • Ceramic Pin + Open
  • Ceramic Pin + Open
  • Temperature range:
  • 0...60 °C
  • 0...60 °C
  • Range:
  • 2 - 12 pH
  • 2 - 12 pH
  • Sensor type:
  • Standard
  • Standard
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Type
    Ø
    Longueur
    Cable Length
    Connecteur
    Fill Solution
    Body Material
    Availability
    Price
  • IntelliCal™ PHC108 (puncture tip)
    Glass
    6 mm
    182 mm
    1 m
    HQD specific
    Non-refillable polymer gel
    Stainless steel
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    10146-090
    Junction
    Ceramic Pin + Open
    Temperature range
    0...60 °C
    Range
    2 - 12 pH
    Sensor type
    Standard
  • IntelliCal™ PHC108 (puncture tip)
    Glass
    6 mm
    182 mm
    1 m
    HQD specific
    Non-refillable polymer gel
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,567.77
    $1,567.77
    Each
       $1,567.77
    Supplier Number: PHC10801
    Spécifications:
    More
    Specifications
    Cat. No.
    CA10146-090
    Junction
    Ceramic Pin + Open
    Temperature range
    0...60 °C
    Range
    2 - 12 pH
    Sensor type
    Standard