Molybdenum Standard Solution, 1000 mg/L as Mo (NIST), 100 mL, Hach
Catalog # CA11016-680
Supplier: Hach
CAS Number:
Specifications
- Concentration:1000 mg/L Mo
- Size:100 ml
- Cat. no.:CA11016-680
- Supplier No.:1418642
- Parameters:Molybdenum as Mo (NIST)
Specifications
About this item
For accuracy checks in molybdenum determinations.