SP Scientific miVac Freeze Drying Flask Kits and Components, Genevac
Supplier: Genevac
Certificates
About this item
The miVac SpeedTrap can be used in conjunction with a Super Vacuum pump.
- Can be used with stand-alone freeze drying accessory kit or add-on flask kits
- Freeze up to 250 ml of water or other suitable solvents
A pressure controller is recommended when samples will be freeze dried using the concentrator to assure optimal vacuum level for your process.
Samples can be placed in a rotor in a miVac concentrator or a range of flask and vial accessories are available to allow the SpeedTrap to directly accept pre-frozen samples for drying.
Ordering information: Includes flask with cap and connection tube. Must be ordered with the freeze drying accessory kit.
Frequently Bought Together