Akro-Grid Dividable Boxes, Akro-Mils
À propos de cet article
Akro-Grid Dividable Boxes are ideal for work-in-process transfer applications
- Divider slots allow users to create hundreds of compartment sizes
- Dividers (sold separately) allow subdivision of the containers to a 1.125" square compartment size
- Made in the USA
Boxes resist most solvents and chemicals. Strong stacking rims and external ribbing provide added strength. Optional snap-on lids keep contents secure and dust free. Dividers are sold separately.
Use dividers to create custom sized storage compartments as small as 1.125".
Informations sur les accessoires: Optional Accessory include Clip-On Tote Label Holder clip on to Akro-Grid Dividable Containers to provide a highly visible label for parts tracking and barcode scanning.
20 Options Available Below
Product Details & Documents
Akro-Grid Dividable Boxes are ideal for work-in-process transfer applications
- Divider slots allow users to create hundreds of compartment sizes
- Dividers (sold separately) allow subdivision of the containers to a 1.125" square compartment size
- Made in the USA
Boxes resist most solvents and chemicals. Strong stacking rims and external ribbing provide added strength. Optional snap-on lids keep contents secure and dust free. Dividers are sold separately.
Use dividers to create custom sized storage compartments as small as 1.125".
Informations sur les accessoires: Optional Accessory include Clip-On Tote Label Holder clip on to Akro-Grid Dividable Containers to provide a highly visible label for parts tracking and barcode scanning.
Product Family Options
Informations sur le produit
- CapacitéColorDimensionsAvailabilityPrice
- Supplier Number: 33105BLUE
- Supplier Number: 33105GREY
- Supplier Number: 33105RED
- Supplier Number: 33162RED
- Supplier Number: 33164BLUE
- Supplier Number: 33164GREY
- Supplier Number: 33164RED
- 35 lbs.Blue10.875×16.5×6" (W×L×H)Discontinued-Supplier Number: -
- Supplier Number: 33166GREY
- Supplier Number: 33166RED
- Supplier Number: 33168BLUE
- Supplier Number: 33168GREY
- 50 lbs.Blue17.375×22.375×10" (W×L×H)Discontinued-Supplier Number: -
- 50 lbs.Grey17.375×22.375×10" (W×L×H)Discontinued-Supplier Number: -
- Supplier Number: 33220RED
- 30 lbs.Blue17.375×22.5×3" (W×L×H)Discontinued-Supplier Number: -
- 30 lbs.Red17.375×22.5×3" (W×L×H)Discontinued-Supplier Number: -
- Supplier Number: 33224BLUE
- Supplier Number: 33224GREY
- Supplier Number: 33224RED
- Supplier Number: 33226BLUE
- Supplier Number: 33226GREY
- Supplier Number: 33226RED
- Supplier Number: 33228GREY
- Supplier Number: 33228RED
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Label Holder for 3.75×5" Labels, Clear
Informations sur le produit
- Catalog #AvailabilityPrice
- Clear Lid for Akro-Grid Boxs 33103/33105Clear Lid for Akro-Grid Boxs 33103/3310575854-454Log in pour la disponibilitéCase of 10 - $254.37
Informations sur le produit
- Catalog #AvailabilityPrice
- Clear Lid for Akro-Grid Boxs 33162/33164/33165/33166/33168Clear Lid for Akro-Grid Boxs 33162/33164/33165/33166/3316875854-458Log in pour la disponibilitéCase of 4 - $153.47
Informations sur le produit
- Catalog #AvailabilityPrice
- Short Divider for Akro-Grid Box 33105
Informations sur le produit
- Catalog #AvailabilityPrice
- Short Divider for Akro-Grid Box 33164
Informations sur le produit
- Catalog #AvailabilityPrice
- Short Divider for Akro-Grid Box 33166
Informations sur le produit
- Catalog #AvailabilityPrice
- Short Divider for Akro-Grid Box 33168
Informations sur le produit
- Catalog #AvailabilityPrice
- Long Divider for Akro-Grid Box 33166
Informations sur le produit
- Catalog #AvailabilityPrice
- Long Divider for Akro-Grid Box 33168
Informations sur le produit
- Catalog #AvailabilityPrice
- Long Divider for Akro-Grid Box 33220
Informations sur le produit
- Catalog #AvailabilityPrice
- Long Divider for Akro-Grid Box 33224
Informations sur le produit
- Catalog #AvailabilityPrice
- Long Divider for Akro-Grid Box 33226
Informations sur le produit
- Catalog #AvailabilityPrice
- Short Divider for Akro-Grid Box 33224
Informations sur le produit
- Catalog #AvailabilityPrice
- Long Divider for Akro-Grid Box 33228
Informations sur le produit
- Catalog #AvailabilityPrice
- Short Divider for Akro-Grid Box 33226
Informations sur le produit
- Catalog #AvailabilityPrice
- Short Divider for Akro-Grid Box 33228
Informations sur le produit
- Catalog #AvailabilityPrice
- Long Divider for Akro-Grid Box 33105
Informations sur le produit
- Catalog #AvailabilityPrice
- Long Divider for Akro-Grid Box 33164