Dissolution Vessels and Centering System Accessories, Quality Lab Accessories
 :   QLA
Dissolution Vessels and Centering System Accessories, Quality Lab Accessories
 :  QLA
Dissolution Vessels and Centering System Accessories, Quality Lab Accessories
Dissolution Vessels and Centering System Accessories, Quality Lab Accessories
QLA manufactures a wide selection of Precision Molded and Standard Glass Vessels for most dissolution testers.
  • All glass vessels are made of borosilicate glass in the USA
  • Vessels are available in clear and amber
  • UltraCenter Precision, Standard and Apex Vessels available with both glass and plastic rims
Good dissolution vessels are critical in obtaining correct and consistent results
UltraCenter Precision Vessels are manufactured using a unique molded process which provides a precise and consistent inside diameter and spherical radius resulting in more reproducible dissolution test results. They are compatible with most dissolution baths. QLA's molded Apex vessels are patent pending.
We have the solution if you are having problems with vessel centering. The QLA AccuCenter Bracket provides ultimate centering accuracy on most dissolution baths (both old and new). It is adjustable and designed to center the vessel from the inside diameter and not the outside. When used with our UltaCenter Precision Vessels with glass or plastic rims, extreme centering accuracy can be achieved. Note: Standard glass vessels cannot be used in the AccuCenter Bracket.
Plastic Rimmed Vessels were designed to help minimize glass rim breakage that can occur in the lab. All QLA plastic rim vessels are dishwasher safe.
All vessels meet the physical dimensions of the USP and have a hemispherical bottom which should be perfectly rounded with no obvious defects. QLA glass vessels are acid etched, or laser marked on plastic rim, with a unique serial number and come with a Certificate of Conformance (COC), where applicable.
 : All vessels are supplied with a Certificate of Conformance (COC) stating that they are USP compliant, where applicable, and that they meet machine manufacturer specifications. A Certificate of Analysis (COA) is available at an extra cost.
Product Family Options
1000mL UltraCenter Precision Molded Glass Vessels
  • Désignation
    Volume
  • Clear UltraCenter Precision Glass Vessel, Serialized
    1000 ml
    Each
    Each - $532.85
    $532.85
    Each
       $532.85
     : ASTM900-UN
  • Amber UltraCenter Precision Glass Vessel, Serialized
    1000 ml
    Each
    Each - $676.43
    $676.43
    Each
       $676.43
     : ASTM900-UNAM
  • Clear UltraCenter Precision Glass Vessel with Plastic Rim, Serialized
    1000 ml
    Each
    Each - $647.71
    $647.71
    Each
       $647.71
     : ASTM900-UNP
  • Amber UltraCenter Precision Glass Vessel with Plastic Rim, Serialized
    1000 ml
    Each
    Each - $727.47
    $727.47
    Each
       $727.47
     : ASTM900-AUNP
  • Clear UltraCenter Precision Glass Vessel for TruCenter, No Magnetic Collar, Serialized
    1000 ml
    Each
    Each - $532.85
    $532.85
    Each
       $532.85
     : ASTM900-VK
  • Amber UltraCenter Precision Glass Vessel for TruCenter, No Magnetic Collar, Serialized
    1000 ml
    Each
    Each - $676.43
    $676.43
    Each
       $676.43
     : ASTM900-AVK
1000mL UltraCenter Precision Molded Apex Glass Vessels
  • Désignation
    Volume
  • Clear UltraCenter Precision Glass Apex Vessel with Plastic Rim, Serialized
    1000 ml
    Each
    Each - $754.62
    $754.62
    Each
       $754.62
     : ASTM9PK-UNP
  • Amber UltraCenter Precision Glass Apex Vessel with Plastic Rim, Serialized
    1000 ml
    Each
    Each - $800.87
    $800.87
    Each
       $800.87
     : ASTM9PK-AUNP
500mL Standard Glass Vessels
  • Désignation
    Volume
  • Clear Glass Vessel for Agilent/VanKel/Varian
    500 ml
    Each
    Each - $285.58
    $285.58
    Each
       $285.58
     : GLA500-01
  • Amber Glass Vessel for Agilent/VanKel/Varian
    500 ml
    Each
    Each - $453.08
    $453.08
    Each
       $453.08
     : GLA500-AV
  • Clear Glass Vessel for Distek, No Ring
    500 ml
    Each
    Each - $279.19
    $279.19
    Each
       $279.19
     : GLA500-DK
  • Amber Glass Vessel for Distek, No Ring
    500 ml
     : -
  • Clear Glass Vessel for Hanson SR8-Plus
    500 ml
    Each
    Each - $285.58
    $285.58
    Each
       $285.58
     : GLA500-HR
1000mL Standard Glass Vessels
  • Désignation
    Volume
  • Clear Glass Vessel for Agilent/VanKel/Varian, Serialized
    1000 ml
    Each
    Each - $264.84
    $264.84
    Each
       $264.84
     : GLA900-01
  • Amber Glass Vessel for Agilent/VanKel/Varian, Serialized
    1000 ml
    Each
    Each - $390.87
    $390.87
    Each
       $390.87
     : GLA900-AV
  • Clear Glass Vessel for TruCenter, No Magnetic Collar, Serialized
    1000 ml
    Each
    Each - $371.73
    $371.73
    Each
       $371.73
     : GLA900-VK
  • Clear Glass Vessel for Distek, No Ring, Serialized
    1000 ml
    Each
    Each - $468.23
    $468.23
    Each
       $468.23
     : GLA900-DK
  • Clear Premier Glass Vessel for Distek, No Ring, Serialized
    1000 ml
    Each
    Each - $681.85
    $681.85
    Each
       $681.85
     : GLA900-DKP
  • Amber Premier Glass Vessel for Distek, No Ring, Serialized
    1000 ml
    Each
    Each - $438.56
    $438.56
    Each
       $438.56
     : GLA900-DKAP
  • Amber Glass Vessel for Distek, No Ring, Serialized
    1000 ml
    Each
    Each - $390.87
    $390.87
    Each
       $390.87
     : GLA900-AD
  • Clear Glass Vessel with Acculign Ring for Distek, Serialized
    1000 ml
    Each
    Each - $298.34
    $298.34
    Each
       $298.34
     : GLA9CR-DK
  • Amber Glass Vessel with Acculign Ring for Distek, Serialized
    1000 ml
    Each
    Each - $413.21
    $413.21
    Each
       $413.21
     : GLA9CR-AD
  • Clear Glass Vessel for Hanson SR8-Plus, Serialized
    1000 ml
    Each
    Each - $245.69
    $245.69
    Each
       $245.69
     : GLA900-HR
  • Amber Glass Vessel for Hanson SR8-Plus, Serialized
    1000 ml
    Each
    Each - $390.87
    $390.87
    Each
       $390.87
     : GLA900-AH
  • Clear Glass Vessel for Erweka, Serialized
    1000 ml
    Each
    Each - $264.84
    $264.84
    Each
       $264.84
     : GLA900-EW
  • Amber Glass Vessel for Erweka, Serialized
    1000 ml
    Each
    Each - $619.30
    $619.30
    Each
       $619.30
     : GLA900-AEW
  • Clear Glass Vessel for Pharmatest, Serialized
    1000 ml
    Each
    Each - $493.50
    $493.50
    Each
       $493.50
     : GLA900-PT
  • Amber Glass Vessel for Pharmatest, Serialized
    1000 ml
    Each
    Each - $390.87
    $390.87
    Each
       $390.87
     : GLA900-APT
  • Clear Glass Vessel for Sotax, Serialized
    1000 ml
    Each
    Each - $416.40
    $416.40
    Each
       $416.40
     : GLA900-ST
  • Amber Glass Vessel for Sotax, Serialized
    1000 ml
    Each
    Each - $583.90
    $583.90
    Each
       $583.90
     : GLA900-AST
  • Clear Glass Vessel for Caleva, Serialized
    1000 ml
    Each
    Each - $264.84
    $264.84
    Each
       $264.84
     : GLA900-CA
  • Amber Glass Vessel for Caleva, Serialized
    1000 ml
    Each
    Each - $568.47
    $568.47
    Each
       $568.47
     : GLA900-AC
  • Clear Glass Vessel with Centering Ring for Zymark/Multidose, Serialized
    1000 ml
    Each
    Each - $279.19
    $279.19
    Each
       $279.19
     : GLA9CR-ZM
  • Amber Glass Vessel with Centering Ring for Zymark/Multidose, Serialized
    1000 ml
    Each
    Each - $561.56
    $561.56
    Each
       $561.56
     : GLA9CR-AZM
  • Clear Glass Vessel with Plastic Rim, Serialized
    1000 ml
    Each
    Each - $379.71
    $379.71
    Each
       $379.71
     : GLA900-UNP
  • Amber Glass Vessel with Plastic Rim, Serialized
    1000 ml
    Each
    Each - $454.68
    $454.68
    Each
       $454.68
     : GLA900-AUNP
1000mL Standard Glass Apex Vessels
  • Désignation
    Volume
  • Clear Glass Apex Vessel for Agilent/VanKel/Varian, Serialized
    1000 ml
    Each
    Each - $339.81
    $339.81
    Each
       $339.81
     : GLA9PK-01
  • Amber Glass Apex Vessel for Agilent/VanKel/Varian, Serialized
    1000 ml
    Each
    Each - $504.12
    $504.12
    Each
       $504.12
     : GLA9PK-AV
  • Clear Glass Apex Vessel for TruCenter, No Magnetic Collar, Serialized
    1000 ml
    Each
    Each - $1,009.42
    $1,009.42
    Each
       $1,009.42
     : GLA9PK-VK
  • Clear Glass Apex Vessel with Acculign Ring for Distek, Serialized
    1000 ml
    Each
    Each - $595.72
    $595.72
    Each
       $595.72
     : GLA9PK-DK
  • Clear Glass Apex Vessel with Ring for Hanson SR8-Plus, Serialized
    1000 ml
    Each
    Each - $602.35
    $602.35
    Each
       $602.35
     : GLA9PK-HR
  • Amber Glass Apex Vessel with Ring for Hanson SR8-Plus, Serialized
    1000 ml
    Each
    Each - $504.12
    $504.12
    Each
       $504.12
     : GLA9PK-AH
  • Clear Glass Apex Vessel with Plastic Rim, Serialized
    1000 ml
    Each
    Each - $489.78
    $489.78
    Each
       $489.78
     : GLA9PK-UNP
  • Amber Glass Apex Vessel with Plastic Rim, Serialized
    1000 ml
    Each
    Each - $618.99
    $618.99
    Each
       $618.99
     : GLA9PK-AUNP
2000mL Standard Glass Vessels
  • Désignation
    Volume
  • Clear Glass Vessel for Agilent/VanKel/Varian, Serialized
    2000 ml
    Each
    Each - $510.51
    $510.51
    Each
       $510.51
     : GLA02K-01
  • Clear Glass Vessel for Distek, No Ring, Serialized
    2000 ml
    Each
    Each - $445.10
    $445.10
    Each
       $445.10
     : GLA02K-DK
  • Amber Glass Vessel for Distek, No Ring, Serialized
    2000 ml
    Each
    Each - $574.33
    $574.33
    Each
       $574.33
     : GLA02K-AD
  • Clear Glass Vessel for Hanson SR8-Plus, Serialized
    2000 ml
    Each
    Each - $510.51
    $510.51
    Each
       $510.51
     : GLA02K-HR
  • Clear Glass Vessel for Erweka, Serialized
    2000 ml
    Each
    Each - $848.62
    $848.62
    Each
       $848.62
     : GLA02K-EW
1000mL Plastic Vessels
  • Désignation
    Volume
  • Clear Plastic Vessel with Feet for Agilent/VanKel/Varian
    1000 ml
    Each
    Each - $270.19
    $270.19
    Each
       $270.19
     : PLA900-01
  • Amber Plastic Vessel with Feet for Agilent/VanKel/Varian
    1000 ml
    Each
    Each - $443.51
    $443.51
    Each
       $443.51
     : PLA900-AV
  • Clear Plastic Vessel with Feet for Distek
    1000 ml
    Each
    Each - $245.69
    $245.69
    Each
       $245.69
     : PLA900-DK
  • Amber Plastic Vessel with Feet for Distek
    1000 ml
    Each
    Each - $443.51
    $443.51
    Each
       $443.51
     : PLA900-AD
  • Clear Plastic Vessel with Feet for Hanson
    1000 ml
    Each
    Each - $270.19
    $270.19
    Each
       $270.19
     : PLA900-HR
  • Amber Plastic Vessel with Feet for Hanson
    1000 ml
    Each
    Each - $443.51
    $443.51
    Each
       $443.51
     : PLA900-AH
  • Clear Plastic Vessel with Feet for Erweka
    1000 ml
    Each
    Each - $440.25
    $440.25
    Each
       $440.25
     : PLA900-EW
  • Clear Plastic Vessel with Feet for Pharmatest
    1000 ml
    Each
    Each - $270.19
    $270.19
    Each
       $270.19
     : PLA900-PT
  • Amber Plastic Vessel with Feet for Pharmatest
    1000 ml
    Each
    Each - $443.51
    $443.51
    Each
       $443.51
     : PLA900-AP
Vessel Holders
  • Désignation
    Volume
  • 6 Position Vessel Stand, Universal
    Each
    Each - $548.80
    $548.80
    Each
       $548.80
     : VESSTD-06H
  • 8 Position Vessel Stand, Universal
    Each
    Each - $999.06
    $999.06
    Each
       $999.06
     : VESSTD-08H
Accessories
  •  
    Description
  • 11010-242
    Vessel Centering Bracket (EaseAlign) with 6 Replaceable Pins for Legacy VanKel Baths
    Each - $174.11
  •  
    Description
  • 11010-244
    Vessel Centering Bracket (EaseAlign) Repair Kit (Set/12)
    Each - $78.19
  •  
    Description
  • 11010-246
    Vessel Centering Bracket Peg
    Each - $42.69
  •  
    Description
  • 11010-170
    Zymark/MultiDose Centering Ring
    Each - $19.72
  •  
    Description
  • 11010-460
    Acculign Plastic Vessel Centering Ring for Distek Baths
    Each - $33.17
  •  
    Description
  • 11010-458
    Zymark/Multidose Vessel Retainer
    Each - $220.16
  •  
    Description
  • 11010-510
    Vessel Retainer Assembly for Distek 2100 and 6300 baths
    Each - $19.49
  •  
    Description
  • 75849-090
    AccuCenter Bracket for Distek 2100 Baths includes hardware
    Each - $276.50
  •  
    Description
  • 75848-944
    Vessel Clamp Knob for 708-DS baths
    Pack of 2 - $82.93
  •  
    Description
  • 75849-092
    AccuCenter Bracket for Distek 2500 Baths includes hardware
    Each - $287.16
  •  
    Description
  • 75849-096
    AccuCenter Bracket for Hanson SR8-Plus Baths includes hardware
    Each - $287.16
  •  
    Description
  • 75849-266
    Vessel Hold Down Ring for UltraCenter Vessels on Erweka DT Baths
    Each - $135.61
  •  
    Description
  • 75849-098
    AccuCenter Bracket for Legacy VanKel and Agilent 708-DS Baths includes hardware
    Each - $287.16