Screw Caps for Tubes, Watson Bio Lab
Coloured screw caps for use with test tubes.
- Resistant to the pressure and freeze
- O-ring made of silicone rubber is resistant to pressure
Suitable for storage in liquid nitrogen (vapor phase) or in deep freezers.
Frequently Bought Together