Synthware® Volumetric Flasks, Heavy Wall, Class A
Class A borosilicate glass volumetric flask.
- Class A
- Heavy wall borosilicate glass construction
- Glass or PTFE stopper
Synthware volumtric flasks are perfect fro precise measurment of liquids in the lab.
Frequently Bought Together