ContactUsLinkComponent
Microscale Slide, Watson Bio Lab
Microscale Slide, Watson Bio Lab
Catalog # 75982-988
Supplier:  Watson Bio Lab
undefined
Microscale Slide, Watson Bio Lab
Catalog # 75982-988
Supplier:  Watson Bio Lab
Supplier Number:  177-401C
Specifications
  • Description:
    Microscale slide
  • Cat. no.:
    75982-988
Specifications
About this item
This polystyrene slide has smooth scale lines.
  • Accurate Linearity
  • Inexpensive
  • Polystyrene
  • Smooth Scale Lines
Users can apply the sample directly on the Microscale when observing with a microscope
Please replace with a new slide after using several times and if it is hard to see due to dirt and scratch.
The economic slide is less expensive than a traditional glass slide.