ContactUsLinkComponent
Ward's® Essentials Chemiluminescence Lab Kit
  470225-518
undefined
Ward's® Essentials Chemiluminescence Lab Kit
  470225-518
 :  AD-WE2502-KIT
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Chemiluminescence Lab Kit
  • No. of Lab Groups:
    15
  • Cat. no.:
    470225-518
  • Supplier No.:
    AD-WE2502-KIT
Create a "cool light"!
  • Hands on lab activity
  • Kit aligned to Next Generation Science Standards MS-PS1-2
  • Instructions and materials included for 15 groups
  • Middle school
In this lab, students will create "cool light" as the result of the oxidation of a chemiluminescent compound. Students will use luminol to mix with an oxidizer and create a chemiluminescent reaction.