Ward's® Essentials Catapult Kit | Avantor
Ward's® Essentials Catapult Kit
Catapult Kit, 1
Catalog # 470225-378
Supplier:   Wards
Fournisseur #:  VWRGP00007
Ward's® Essentials Catapult Kit
Catalog # 470225-378
Supplier:  Wards
Ward's® Essentials Catapult Kit
Ward's® Essentials Catapult Kit
Catapult Kit, 1
Price
71,09 $
Log in pour la disponibilité
Kit of 1
Product Details & Documents
Play with the physics of a catapult to learn about levers, force, acceleration and much more!
  • Hands on lab activity
  • Kit aligned to Next Generation Science Standards MS-PS2-2
  • Instructions and materials included for 1 group
  • Middle school
Explore force, acceleration, Hooke's law, and parabolic trajectories by building and testing a fully-functional catapult. Contains catapult base, sides, lever arm, rubber bands, screws, dowels, hook, and ping-pong ball.
Spécifications
  • Désignation:
    Catapult Kit
  • No. of Lab Groups:
    1