Hyper Filter Tips, Watson Bio Lab
These filtered pipette tips feature special hydrophobic porosity filters with fluorocarbon resin that includes PTFE.
- Sterilized
- Filters Contain fluorocarbon
- Resin with PTFE
- Dense, hydrophobic porosity filters
- Excellent chemical resistance
- Easy sample collection
These filter tips prevent sample overflow when aspiration occurs due to human error or contamination occurs due to aerosol and DNA.
The filter tips prevent the sample overflow the minute after the solution is absorbed.
: Empty racks are available separately.
Frequently Bought Together