NEXTY Tip Racks, Watson Bio Lab
Fournisseur: Watson Bio Lab
Certificats
À propos de cet article
Empty PC racks for use with NEXTY pipette tips.
- Designed for "durable" and "repeated autoclaving" (repeated autoclaving may cause damages to the rack)
- Stable design: Trapezoid shape allows tips to fit into the rack without holding the rack in the hand
- Non-slip rubber base enhances stability (Removable and not autoclavable)
- Four drain holes on the bottom allows quick drying after autoclaving
- Hinged lid opens wide enough to access the tips even with multi channel pipettes
- One touch operation: Lid can be opened and closed with just one finger
- Lid automatically locks when closed
The lock latch can be colour coded. Select from 3 color options for easy identification (3 pieces are included).
Frequently Bought Together