NEXTY Fixed Volume Pipettors, Watson Bio Lab
Supplier: Avantor
Certificates
About this item
The NEXTY fixed volume pipettor has an equal volume adjustment mechanism so you can customize your original fixed volume pipette.
- Mushroom-shaped push button is designed to repetitive stress on the thumb
- Unique design of the eject button provides for easy and comfortable ejecting and dispensing
- Nozzle cone and eject cone can easily be removed for cleaning, they are also autoclavable
The NEXTY pipettes have passed the Half Million stroke test. The spring system and PBT internal plunger (which is highly resistant to chemicals and samples) means these units always have a smooth and precise stroke.
Frequently Bought Together