Order Entry
Canada
ContactUsLinkComponent
Silica Standard Solution, 50 mg/ml as SiO₂ (NIST), 200 ml, Hach
Silica Standard Solution, 50 mg/ml as SiO₂ (NIST), 200 ml, Hach
Catalog # CA11016-526
Supplier:  Hach
CAS Number:  
undefined
Silica Standard Solution, 50 mg/ml as SiO₂ (NIST), 200 ml, Hach
Catalog # CA11016-526
Supplier:  Hach
Supplier Number:  111729
CAS Number:  

Specifications

  • Concentration:
    50 mg/L SiO₂
  • Size:
    200 ml
  • Cat. no.:
    CA11016-526
  • Supplier No.:
    111729
  • Parameters:
    Sulfate as SiO₂ (NIST)

Specifications

About this item

For accuracy checks in silica determinations.