To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:Micro-Cut Mini-Tischler Biopsy Punch
- Length:20.6 cm (8")
- Material:Stainless steel
- Tip Configuration:Semi Elliptical/Semi Oval Bite with 1 Lower Tooth
- Curvature:Straight
- Cat. no.:10194-152
- Shape:Rectangular
- Supplier No.:90-8945
- Tip Type:With locking tip
Sklar's Micro-Cut Mini Tischler Biopsy Punch Forceps are a non-ratcheted, double spring instrument used to cut tissue samples in gynecologic procedures.
This product is straight with an 8 inch shaft and 2.3mm x 4.2mm rectangular bite. It features a single, lower tooth used to grasp difficult to reach tissue and a locking mechanism above the horn on the handle so the jaws can lock the tissue sample into place while the instrument is removed from the cervical canal. This reduces the risk of losing the tissue sample as the instrument is withdrawn. This product has a pistol style handle with one large ring for multiple fingers. Micro-Cut biopsy punches are identified by gold handles.