Silica Standard Solution, 1 mg/ml as SiO₂ (NIST), 500 ml, Hach
CA1106-49
: Hach
:
- Concentration:1 mg/L SiO₂
- Size:500 ml
- Cat. no.:CA1106-49
- Supplier No.:1106-49
- Parameters:Silica as SiO2 (NIST)
For accuracy checks in silica determinations.