ContactUsLinkComponent
Bulk uTIP™ Pipette Tips, Biotix
Bulk uTIP™ Pipette Tips, Biotix
Supplier:  Biotix
   Sustainable Options Available
Certificates
undefined
Bulk uTIP™ Pipette Tips, Biotix
Supplier:  Biotix
   Sustainable Options Available
Certificates
Specifications
  • Catalog No:
  • 75982-124
  • 75982-126
  • 75982-128
  • 75982-130
  • Environmentally Preferable:
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Environmental certification:
  • FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%
  • FSC-STD-40-004 V3-1 and FSC-C1544854/30/2025; Bulk tips reduce plastic waste by 95%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%