Vernier® Go Direct™ pH Sensor
New Product
Vernier® Go Direct™ pH Sensor
New Product
Certificates
Vernier® Go Direct™ pH Sensor
Vernier® Go Direct™ pH Sensor
3 Options Available Below
Product Details & Documents
A general-purpose pH sensor used to monitor pH of aqueous solutions.
  • Conduct acid-base titrations
  • Monitor pH change during chemical reactions
  • Test the pH and alkalinity of bodies of water
  • Investigate household acids and bases
The Go Direct pH Sensor is an important and versatile sensor for lab and field activities alike. It gives students the freedom to explore pH without the inconvenience of wires—no more spilled solutions or dangling wires. The Go Direct pH transmits live readings and captures data in real-time.
This sensor is comprised of a pH electrode connected to a Go Direct Electrode Amplifier via a BNC connector. This design enables you to use the Go Direct Electrode Amplifier with other Vernier electrodes, such as the Flat pH Electrode, ORP Electrode, ISE electrode, or a third-party electrode.
The Go Direct robust rechargeable battery offers long battery life and provides always-ready operation. Monitor battery life directly from our free Graphical Analysis™ 4 app. Should the battery run low, simply connect the sensor to the charging cable and keep on collecting—no interruptions or inconvenience to you or your students.
Our Go Direct™ pH Sensor connects directly to your mobile device, Chromebook, or computer using our free Graphical Analysis™ 4 app—no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth, allowing you to choose the solution best for your classroom or laboratory.
 : Go Direct pH sensor includes Go Direct Electrode amplifier,pH storage solution bottle with pH storage solution, micro USB cable, quick-start guide.
Go Direct pH Teacher Pack includes eight Go Direct pH Sensors, eight pH storage solution bottles with pH storage solution, charge station, eight micro USB cables, Quick-start guide.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Go Direct™ pH sensor
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $214.09
    $214.09
    Each
       $214.09
    Supplier Number: GDX-PH
    Spécifications:
    New Product
    New Product
  • Go Direct™ pH replacement electrode
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $90.09
    $90.09
    Each
       $90.09
    Supplier Number: GDX-PH-BNC
    Spécifications:
    New Product
    New Product
  • Go Direct™ pH teacher pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 8
    Pack of 8 - $1,894.09
    $1,894.09
    Pack of 8
       $1,894.09
    Supplier Number: GDX-PH-TP
    Spécifications:
    New Product
    New Product
Storage Solution
Informations sur le produit
  • Catalog #
    Désignation
    Availability
    Price
  • Vernier pH storage solution, 500 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $52.09
    $52.09
    Each
       $52.09