Dissolution Capsule Sinkers for Apparatus 2, Quality Lab Accessories
Supplier: QLA
Certificates
About this item
QLA offers the widest variety of Capsule Sinkers in the Dissolution Industry.
- Made of high quality 316 stainless steel or PTFE coated over music wire
- All sinkers are made in the USA
- Custom sinkers are available upon request
Home-made sinkers tend not to be reproducible and so commercially available sinkers are quite popular
USP (711) states: " A small, loose piece of non-reactive material such as not more than a few turns of wire helix may be attached to dosage units that would otherwise float. Other validated sinker devices may be used."
The choice of sinker will depend on the tablet or capsule. The sinker should have minimal contact with the doasge form as this can affect the dissolution rate. The sinker should be heavy enough to sink the tablet/capsule to the bottom of the vessel. There are many varieties of sinkers including spiral, basket, pronged and spring style.
Frequently Bought Together