HPLC Columns, YMC-Pack Diol-SEC | Avantor
HPLC Columns, YMC-Pack Diol-SEC
Supplier:   YMC America,Inc
HPLC Columns, YMC-Pack Diol-SEC
Supplier:  YMC America,Inc
Certificates
28 Options Available Below
Product Details & Documents
Diol-SEC is a size exclusion chromatography column utilizing a dihydroxypropyl stationary phase bonded to a silica gel base and useful for molecular weight determination of proteins and sugars.
  • Silica-based column with high mechanical strength
  • Very low nonspecific adsorption of proteins
  • More accurate quantitation of monoclonal antibodies with two products
  • Provides improvement in resolution and throughput on size exclusion chromatography
Diol-120, 200, and 300 are suitable for separation or molecular weight determination of proteins with molecular weights of 10000 to several hundred thousand. Diol-60 is the most suitable for separation of peptides of oligosaccharides whose molecular weights are 10000 or less.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 89216-696
  • 89198-934
  • 89216-682
  • 89198-930
  • 89216-686
  • 89216-690
  • 89216-698
  • 97030-794
  • 97030-822
  • 97030-790
  • 97030-752
  • 97030-754
  • 97030-774
  • 97030-796
  • 97030-798
  • 97030-824
  • 97030-826
  • 97030-758
  • 97030-760
  • 97030-772
  • 97030-778
  • 97030-802
  • 97030-804
  • 97030-830
  • 97030-832
  • 97030-792
  • 97030-800
  • 97030-806
  • pH range:
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
  • 5 - 7.5
Spécifications
Product Family Options
Informations sur le produit
  • Type de colonne
    Phase
    Taille de pore
    Column Hardware
    Granulométrie
    Ø int.
    Longueur
    Packing Material
    Availability
    Price
  • Analytique
    Diol-SEC
    300 Å
    Acier inoxydable
    5 µm
    4.00 mm
    50 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,287.52
    $1,287.52
    Each
       $1,287.52
    Supplier Number: DL30S05-0504WT
    Spécifications:
    More
    Specifications
    Cat. No.
    89216-696
    pH range
    5 - 7.5
  • Analytique
    Diol-SEC
    60 Å
    Acier inoxydable
    5 µm
    4.60 mm
    250 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,953.28
    $2,953.28
    Each
       $2,953.28
    Supplier Number: DL06S05-2546WT
    Spécifications:
    More
    Specifications
    Cat. No.
    89198-934
    pH range
    5 - 7.5
  • Analytique
    Diol-SEC
    60 Å
    Acier inoxydable
    5 µm
    4.60 mm
    300 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,337.54
    $3,337.54
    Each
       $3,337.54
    Supplier Number: DL06S05-3046WT
    Spécifications:
    More
    Specifications
    Cat. No.
    89216-682
    pH range
    5 - 7.5
  • Analytique
    Diol-SEC
    120 Å
    Acier inoxydable
    5 µm
    4.60 mm
    250 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,925.03
    $2,925.03
    Each
       $2,925.03
    Supplier Number: DL12S05-2546WT
    Spécifications:
    More
    Specifications
    Cat. No.
    89198-930
    pH range
    5 - 7.5
  • Analytique
    Diol-SEC
    120 Å
    Acier inoxydable
    5 µm
    4.60 mm
    300 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,337.54
    $3,337.54
    Each
       $3,337.54
    Supplier Number: DL12S05-3046WT
    Spécifications:
    More
    Specifications
    Cat. No.
    89216-686
    pH range
    5 - 7.5
  • Analytique
    Diol-SEC
    200 Å
    Acier inoxydable
    5 µm
    4.60 mm
    300 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,337.54
    $3,337.54
    Each
       $3,337.54
    Supplier Number: DL20S05-3046WT
    Spécifications:
    More
    Specifications
    Cat. No.
    89216-690
    pH range
    5 - 7.5
  • Analytique
    Diol-SEC
    300 Å
    Acier inoxydable
    5 µm
    4.60 mm
    300 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,492.13
    $3,492.13
    Each
       $3,492.13
    Supplier Number: DL30S05-3046WT
    Spécifications:
    More
    Specifications
    Cat. No.
    89216-698
    pH range
    5 - 7.5
  • Analytique
    Diol-SEC
    200 Å
    Acier inoxydable
    5 µm
    6.00 mm
    300 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,218.22
    $4,218.22
    Each
       $4,218.22
    Supplier Number: DL20S05-3006WT
    Spécifications:
    More
    Specifications
    Cat. No.
    97030-794
    pH range
    5 - 7.5
  • Analytique
    Diol-SEC
    300 Å
    Acier inoxydable
    5 µm
    6.00 mm
    300 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,218.22
    $4,218.22
    Each
       $4,218.22
    Supplier Number: DL30S05-3006WT
    Spécifications:
    More
    Specifications
    Cat. No.
    97030-822
    pH range
    5 - 7.5
  • Colonne de garde
    Diol-SEC
    200 Å
    Acier inoxydable
    5 µm
    8.00 mm
    30 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,124.52
    $2,124.52
    Each
       $2,124.52
    Supplier Number: DL20S05-0308WTG
    Spécifications:
    More
    Specifications
    Cat. No.
    97030-790
    pH range
    5 - 7.5
  • Analytique
    Diol-SEC
    60 Å
    Acier inoxydable
    5 µm
    8.00 mm
    300 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,237.98
    $5,237.98
    Each
       $5,237.98
    Supplier Number: DL06S05-3008FG
    Spécifications:
    More
    Specifications
    Cat. No.
    97030-752
    pH range
    5 - 7.5
  • Analytique
    Diol-SEC
    60 Å
    Acier inoxydable
    5 µm
    8.00 mm
    300 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,389.30
    $4,389.30
    Each
       $4,389.30
    Supplier Number: DL06S05-3008WT
    Spécifications:
    More
    Specifications
    Cat. No.
    97030-754
    pH range
    5 - 7.5
  • Analytique
    Diol-SEC
    120 Å
    Acier inoxydable
    5 µm
    8.00 mm
    300 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,409.22
    $4,409.22
    Each
       $4,409.22
    Supplier Number: DL12S05-3008WT
    Spécifications:
    More
    Specifications
    Cat. No.
    97030-774
    pH range
    5 - 7.5
  • Analytique
    Diol-SEC
    200 Å
    Verre
    5 µm
    8.00 mm
    300 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,237.98
    $5,237.98
    Each
       $5,237.98
    Supplier Number: DL20S05-3008FG
    Spécifications:
    More
    Specifications
    Cat. No.
    97030-796
    pH range
    5 - 7.5
  • Analytique
    Diol-SEC
    200 Å
    Acier inoxydable
    5 µm
    8.00 mm
    300 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,368.09
    $4,368.09
    Each
       $4,368.09
    Supplier Number: DL20S05-3008WT
    Spécifications:
    More
    Specifications
    Cat. No.
    97030-798
    pH range
    5 - 7.5
  • Analytique
    Diol-SEC
    300 Å
    Acier inoxydable
    5 µm
    8.00 mm
    300 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,237.98
    $5,237.98
    Each
       $5,237.98
    Supplier Number: DL30S05-3008FG
    Spécifications:
    More
    Specifications
    Cat. No.
    97030-824
    pH range
    5 - 7.5
  • Analytique
    Diol-SEC
    300 Å
    Acier inoxydable
    5 µm
    8.00 mm
    300 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,534.02
    $4,534.02
    Each
       $4,534.02
    Supplier Number: DL30S05-3008WT
    Spécifications:
    More
    Specifications
    Cat. No.
    97030-826
    pH range
    5 - 7.5
  • Analytique
    Diol-SEC
    60 Å
    Acier inoxydable
    5 µm
    8.00 mm
    500 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,416.64
    $7,416.64
    Each
       $7,416.64
    Supplier Number: DL06S05-5008FG
    Spécifications:
    More
    Specifications
    Cat. No.
    97030-758
    pH range
    5 - 7.5
  • Analytique
    Diol-SEC
    60 Å
    Acier inoxydable
    5 µm
    8.00 mm
    500 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,621.17
    $6,621.17
    Each
       $6,621.17
    Supplier Number: DL06S05-5008WT
    Spécifications:
    More
    Specifications
    Cat. No.
    97030-760
    pH range
    5 - 7.5
  • Analytique
    Diol-SEC
    120 Å
    Acier inoxydable
    5 µm
    8.00 mm
    300 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,237.98
    $5,237.98
    Each
       $5,237.98
    Supplier Number: DL12S05-3008FG
    Spécifications:
    More
    Specifications
    Cat. No.
    97030-772
    pH range
    5 - 7.5
  • Analytique
    Diol-SEC
    120 Å
    Acier inoxydable
    5 µm
    8.00 mm
    500 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,416.64
    $7,416.64
    Each
       $7,416.64
    Supplier Number: DL12S05-5008FG
    Spécifications:
    More
    Specifications
    Cat. No.
    97030-778
    pH range
    5 - 7.5
  • Analytique
    Diol-SEC
    200 Å
    Verre
    5 µm
    8.00 mm
    500 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,416.64
    $7,416.64
    Each
       $7,416.64
    Supplier Number: DL20S05-5008FG
    Spécifications:
    More
    Specifications
    Cat. No.
    97030-802
    pH range
    5 - 7.5
  • Analytique
    Diol-SEC
    200 Å
    Acier inoxydable
    5 µm
    8.00 mm
    500 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,568.26
    $6,568.26
    Each
       $6,568.26
    Supplier Number: DL20S05-5008WT
    Spécifications:
    More
    Specifications
    Cat. No.
    97030-804
    pH range
    5 - 7.5
  • Analytique
    Diol-SEC
    300 Å
    Acier inoxydable
    5 µm
    8.00 mm
    500 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,416.64
    $7,416.64
    Each
       $7,416.64
    Supplier Number: DL30S05-5008FG
    Spécifications:
    More
    Specifications
    Cat. No.
    97030-830
    pH range
    5 - 7.5
  • Analytique
    Diol-SEC
    300 Å
    Acier inoxydable
    5 µm
    8.00 mm
    500 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,468.67
    $6,468.67
    Each
       $6,468.67
    Supplier Number: DL30S05-5008WT
    Spécifications:
    More
    Specifications
    Cat. No.
    97030-832
    pH range
    5 - 7.5
  • Colonne de garde
    Diol-SEC
    200 Å
    Acier inoxydable
    5 µm
    20.00 mm
    50 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,549.34
    $4,549.34
    Each
       $4,549.34
    Supplier Number: DL20S05-0520WTG
    Spécifications:
    More
    Specifications
    Cat. No.
    97030-792
    pH range
    5 - 7.5
  • Semi-préparative
    Diol-SEC
    200 Å
    Acier inoxydable
    5 µm
    20.00 mm
    300 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $19,707.21
    $19,707.21
    Each
       $19,707.21
    Supplier Number: DL20S05-3020WT
    Spécifications:
    More
    Specifications
    Cat. No.
    97030-800
    pH range
    5 - 7.5
  • Semi-préparative
    Diol-SEC
    200 Å
    Acier inoxydable
    5 µm
    20.00 mm
    500 mm
    Silica
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $29,582.34
    $29,582.34
    Each
       $29,582.34
    Supplier Number: DL20S05-5020WT
    Spécifications:
    More
    Specifications
    Cat. No.
    97030-806
    pH range
    5 - 7.5
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Cartridge Holder, 1.0 – 4.0 mm for 10 mm Length Cartridges
    Cartridge Holder, 1.0 – 4.0 mm for 10 mm Length Cartridges
    89230-180
    Log in pour la disponibilité
    Each - $664.26