HPLC Columns, YMC Carotenoid
Supplier: YMC America,Inc
Certificates
About this item
The Carotenoid C30 stationary phase provides sufficient phase thickness to enhance interaction with long chain molecules therefore, geometric and positional isomers of conjugated double-bonding systems are recognized and resolved.
- These columns is much more hydrophobic and lipophilic
- Even when pure organic eluents are applied, many sample solutes are retained
- Applicable for LC-MS
Carotenoids are found in a variety of natural sources including fruits and vegetables. In addition, carotenoids are considered as potential drugs for cancer intervention and/or prevention. Despite the complexity of carotenoid extracts and the minor shape differences between carotenoid isomers, the separation, identification, and quantitation of these compounds can be achieved using these columns.
Specifications
- Catalog No:
- 89216-678
- 89216-676
- 89216-680
- 89216-670
- 89216-668
- 89216-674
- 89216-672
- 97030-718
- 97030-672
- 97030-716
- 97030-662
- 97030-706
- 97030-704
- 97030-702
- 97030-670
- 97030-714
- 97030-668
- 97030-712
- 97030-666
- 97030-710
- 97030-664
- 97030-708
- 89216-662
- 89216-656
- 89216-660
- 97030-700
- 97030-698
- 89216-666
- 97030-694
- 89216-664
- 97030-660
- 97030-658
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- USP code:
- L62
- L62
- L62
- L62
- L62
- L62
- L62
- L62
- L62
- L62
- L62
- L62
- L62
- L62
- L62
- L62
- L62
- L62
- L62
- L62
- L62
- L62
- L62
- L62
- L62
- L62
- L62
- L62
- L62
- L62