Sample bags available in a variety of materials.
- Customizable
- Accord bags are economical and strong
Micro Por Plus PE bags provide the right combination of properties for the most accurate assay results. Material will handle up to 232 °C (450 °F). Bags are extremely strong and rugged and are inert to most organic chemicals (acids, base, salts). Available drawstring and 5×10 cm (2×4") durable, waterproof tag. Material strength holds up very well over time.
Other materials (Cotton or Canvas), custom sizes and additional options (such as bar-coding) are available on a custom quote basis. Contact your Avantor sales representative for more information.
Spun-bonded polypropylene fabric drains quickly, while retaining fines. Great for collecting large, wet samples. Fibers are evenly distributed to allow for some stretch.