Multirange Electrical Meter
Multirange Electrical Meter
Catalog # 470225-200
Supplier: Shiv Dial Sud & Sons
: PH64250
Jump to:
Product Details & Documents
Design to serve dual purpose as volt meter and ammeter.
- Compact design
- Works as an ammeter and voltmeter
It offers quadruple range and a slide selector is provided to select the required range.
Spécifications
- Désignation:Multirange Electrical Meter
- Measurement range:Volt : 0-100 mV, 1 V, 10 V and 50 V;
Ampere : 0-100 µA, 100 mA, 1 A and 5 A - Cat. No.:470225-200
- Accuracy:±2.5 FSD